missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VAMP7.
Brand: Abnova PAB27200.100ug
This item is not returnable.
View return policy
Specifications
| VAMP7 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| VAMP7 | |
| VAMP7 | |
| A synthetic peptide corresponding to 18 amino acids at N-terminus of human VAMP7. | |
| 100 μg | |
| Primary | |
| At least three isoforms of VAMP7 are known to exist; this antibody will detect the two longest isoforms. This antibody is predicted to not cross-react with other Ras-related proteins. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of VAMP7. | |
| In PBS (0.02% sodium azide) | |
| SYBL1/TI-VAMP/VAMP-7 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 6845 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction