Learn More
UTY, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant UTY.
Brand: Abnova PAB20036.100uL
Description
This gene encodes a protein containing tetratricopeptide repeats which are thought to be involved in protein-protein interactions. This protein is a minor histocompatibility antigen which may induce graft rejection of male stem cell grafts. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: SNSCILLDKCPPPRPPTSPYPPLPKDKLNPPTPSIYLENKRDAFFPPLHQFCTNPKNPVTVIRGLAGALKLDLGLFSTKTLVEANNEHMVEVRTQLLQPADENWDPTGTKKIWRCESNRSHTTISpecifications
| UTY | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant UTY. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp686L12190/UTY1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7404 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry The optimal working dilution should be determined by the end user. | |
| UTY | |
| UTY | |
| Recombinant protein corresponding to amino acids of human UTY. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only