Learn More
UST, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant UST.
Brand: Abnova PAB22603.100uL
Description
Uronyl 2-sulfotransferase transfers sulfate to the 2-position of uronyl residues, such as iduronyl residues in dermatan sulfate and glucuronyl residues in chondroitin sulfate (Kobayashi et al., 1999 [PubMed 10187838]).[supplied by OMIM
Sequence: YNRVGKCGSRTVVLLLRILSEKHGFNLVTSDIHNKTRLTKNEQMELIKNISTAEQPYLFTRHVHFLNFSRFGSpecifications
| UST | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant UST. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| 2OST | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10090 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| UST | |
| UST | |
| Recombinant protein corresponding to amino acids of human UST. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only