missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP6NL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant USP6NL.
Brand: Abnova PAB23523.100uL
This item is not returnable.
View return policy
Description
Sequence: SRQYNHAAANQNSNATSNIRKEFVPKWNKPSDVSATERTAKYTMEGKGRAAHPALAVTVPGPAEVRVSNVRPKMKALDAEDGKRGSTASSpecifications
| USP6NL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant USP6NL. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA0019/RNTRE/TRE2NL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9712 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| USP6NL | |
| USP6NL | |
| Recombinant protein corresponding to amino acids of human USP6NL. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu