Learn More
USP16 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of USP16.
Brand: Abnova PAB4203.400uL
Description
This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
| USP16 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of USP16. | |
| In PBS (0.09% sodium azide) | |
| UBP-M | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 10600 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| USP16 | |
| USP16 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human USP16. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only