Learn More
USH1G, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant USH1G.
Brand: Abnova PAB21685.100uL
Description
This gene encodes a protein that contains three ankyrin domains, a class I PDZ-binding motif and a sterile alpha motif. The encoded protein interacts with harmonin, which is associated with Usher syndrome type 1C. This protein plays a role in the development and maintenance of the auditory and visual systems and functions in the cohesion of hair bundles formed by inner ear sensory cells. Mutations in this gene are associated with Usher syndrome type 1G (USH1G). [provided by RefSeq
Sequence: NIWCLDNDYHTPLDMAAMKGHMECVRYLDSIAAKQSSLNPKLVGKLKDKAFREAERRIRECAKLQRRHHERMERSpecifications
| USH1G | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant USH1G. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| ANKS4A/FLJ33924/SANS | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 124590 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| USH1G | |
| USH1G | |
| Recombinant protein corresponding to amino acids of human USH1G. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only