Learn More
UQCRQ, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant UQCRQ.
Brand: Abnova PAB24512.100uL
Description
This gene encodes a ubiquinone-binding protein of low molecular mass. This protein is a small core-associated protein and a subunit of ubiquinol-cytochrome c reductase complex III, which is part of the mitochondrial respiratory chain. [provided by RefSeq
Sequence: REFGNLTRMRHVISYSLSPFEQRAYPHVFTKGIPNVLRRSpecifications
| UQCRQ | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant UQCRQ. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| QCR8/QP-C/QPC | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 27089 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| UQCRQ | |
| UQCRQ | |
| Recombinant protein corresponding to amino acids of human UQCRQ. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only