Learn More
UPK1B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UPK1B.
Brand: Abnova PAB25730.100ug
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is found in the asymmetrical unit membrane (AUM) where it can form a complex with other transmembrane 4 superfamily proteins. It may play a role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions. The use of alternate polyadenylation sites has been found for this gene. [provided by RefSeq
Specifications
| UPK1B | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| UPK1B | |
| UPK1B | |
| A synthetic peptide corresponding to 17 amino acids at internal region of human UPK1B. | |
| 100 μg | |
| Primary | |
| At least two isoforms of UPK1B are known to exist; this antibody will recognize both isoforms. UPK1B antibody is predicted to not cross-react with other tetraspanin protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of UPK1B. | |
| In PBS (0.02% sodium azide) | |
| TSPAN20/UPIB/UPK1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 7348 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only