Learn More
UFM1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant UFM1.
Brand: Abnova PAB23448.100uL
Description
UFM1 is a ubiquitin-like protein that is conjugated to target proteins by E1-like activating enzyme UBA5 (UBE1DC1; MIM 610552) and E2-like conjugating enzyme UFC1 (MIM 610554) in a manner analogous to ubiquitylation (see UBE2M; MIM 603173) (Komatsu et al., 2004 [PubMed 15071506]).[supplied by OMIM
Sequence: IRAFPTTTPRSLHLFTSSTFLARALPGAFPTGACEERLSVPESTPFTAVLKFAAEEFKVPAATSAIITNDGIGINPAQTAGNVFLKHGSELRIIPRDRVGSCSpecifications
| UFM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant UFM1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| BM-002/C13orf20/bA131P10.1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51569 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| UFM1 | |
| UFM1 | |
| Recombinant protein corresponding to amino acids of human UFM1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only