Learn More
UBR5 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UBR5.
Brand: Abnova PAB27095.100uL
Description
This gene encodes a progestin-induced protein, which belongs to the HECT (homology to E6-AP carboxyl terminus) family. The HECT family proteins function as E3 ubiquitin-protein ligases, targeting specific proteins for ubiquitin-mediated proteolysis. This gene is localized to chromosome 8q22 which is disrupted in a variety of cancers. This gene potentially has a role in regulation of cell proliferation or differentiation. [provided by RefSeq
Specifications
| UBR5 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| UBR5 | |
| UBR5 | |
| A synthetic peptide corresponding to human UBR5. | |
| 100 μL | |
| Primary | |
| UBR5 polyclonal antibody detects endogenous levels of UBR5 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of UBR5. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| DD5/EDD/EDD1/FLJ11310/HYD/KIAA0896/MGC57263 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51366 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only