Learn More
UBE4B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UBE4B.
Brand: Abnova PAB1930.400uL
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. This gene is also the strongest candidate in the neuroblastoma tumor suppressor genes. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
| UBE4B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of UBE4B. | |
| In PBS (0.09% sodium azide) | |
| E4/HDNB1/KIAA0684/UBOX3/UFD2 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 10277 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| UBE4B | |
| UBE4B | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human UBE4B. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only