Learn More
UBE2V1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UBE2V1.
Brand: Abnova PAB12933.100ug
Description
Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein encoded by this gene is located in the nucleus and can cause transcriptional activation of the human FOS proto-oncogene. It is thought to be involved in the control of differentiation by altering cell cycle behavior. Multiple alternatively spliced transcripts encoding different isoforms have been described for this gene. A pseudogene has been identified which is also located on chromosome 20. Co-transcription of this gene and the neighboring upstream gene generates a rare transcript (Kua-UEV), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product. [provided by RefSeq
Specifications
| UBE2V1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of UBE2V1. | |
| In PBS (0.02% sodium azide) | |
| CIR1/CROC-1/CROC1/UBE2V/UEV-1/UEV1/UEV1A | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Anti-UEV1A may also recognize other isoforms of UEV1A. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| UBE2V1 | |
| UBE2V1 | |
| A synthetic peptide corresponding to C-terminus 15 amino acids of human UBE2V1. | |
| RUO | |
| 7335 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only