Learn More
UBE2L3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant UBE2L3.
Brand: Abnova PAB28318.100uL
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to participate in the ubiquitination of p53, c-Fos, and the NFkb precursor p105 in vitro. Several alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: MAASRRLMKELEEIRKCGMKNFRNIQVDEANLLTWQGLIVPDNPPYDKGAFRIEINFPAEYPFKSpecifications
| UBE2L3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant UBE2L3. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| E2-F1/L-UBC/UBCH7/UbcM4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7332 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| UBE2L3 | |
| UBE2L3 | |
| Recombinant protein corresponding to amino acids of recombinant UBE2L3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only