Learn More
UBE2J1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UBE2J1.
Brand: Abnova PAB11288.100ug
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum (ER) and may contribute to quality control ER-associated degradation by the ubiquitin-proteasome system. [provided by RefSeq
Specifications
| UBE2J1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of UBE2J1. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| CGI-76/HSPC153/HSPC205/HSU93243/MGC12555/NCUBE1/Ubc6p | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:1000-1:5000) Western Blot (1:200-1:2000) Immunoprecipitation (1-2 ug) The optimal working dilution should be determined by the end user. | |
| UBE2J1 | |
| UBE2J1 | |
| A synthetic peptide corresponding to N-terminus of human UBE2J1. | |
| RUO | |
| 51465 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only