Learn More
UBE2H, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UBE2H.
Brand: Abnova PAB3899.400uL
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein sequence is 100% identical to the mouse homolog and 98% identical to the frog and zebrafish homologs. Two alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms. [provided by RefSeq
Specifications
| UBE2H | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of UBE2H. | |
| In PBS (0.09% sodium azide) | |
| E2-20K/UBC8/UBCH/UBCH2 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 7328 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| UBE2H | |
| UBE2H | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human UBE2H. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only