missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Turkey IgG whole molecule, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB9477.2mL
This item is not returnable.
View return policy
Specifications
| Rabbit anti-Turkey IgG whole molecule | |
| Polyclonal | |
| Rabbit anti-turkey IgG recognizes turkey IgG whole molecule. This secondary antibody was purified using delipidation and defibrination. | |
| Lyophilized from 10mM sodium phosphate, 250mM NaCl, pH 7.4 (0.01% (w/v) sodium azide) | |
| Turkey IgG whole molecule | |
| 2 mL | |
| Turkey | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against turkey IgG and turkey serum. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction