Learn More
TSSC4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TSSC4.
Brand: Abnova PAB23855.100uL
Description
This gene is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. This gene is located among several imprinted genes; however, this gene, as well as the pan-hematopoietic expression gene (PHEMX), escapes imprinting. This gene may play a role in malignancies and disease that involve this region. [provided by RefSeq
Sequence: TKYSLEDVTEVSEQSNQATALAFLGSQSLAAPTDCVSSFNQDPSSCGEGRVIFTKPVRGVEARHERKRVLGKVGEPGRGGLGNPATDSpecifications
| TSSC4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TSSC4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| TSSC4 | |
| Recombinant protein corresponding to amino acids of human TSSC4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
| TSSC4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10078 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only