Learn More
TSPYL2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TSPYL2.
Brand: Abnova PAB24298.100uL
Description
This gene encodes a member of the testis-specific protein Y-encoded, TSPY-like/SET/nucleosome assembly protein-1 superfamily. The encoded protein is localized to the nucleolus where it functions in chromatin remodeling and as an inhibitor of cell-cycle progression. This protein may play a role in the suppression of tumor growth. [provided by RefSeq
Sequence: NPLRYYLRERGSRIKRKKQEMKKRKTRGRCEVVIMEDAPDYYAVEDIFSEISDIDETIHDIKISDFMETTDYFETTDNEITDINENICDSENPDHNEVPNNETTDNNESADDHETTDNNESADDNNENPEDNNKNTDDSpecifications
| TSPYL2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TSPYL2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CDA1/CINAP/CTCL/DENTT/HRIHFB2216/SE20-4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 64061 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:2500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| TSPYL2 | |
| TSPYL2 | |
| Recombinant protein corresponding to amino acids of human TSPYL2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only