Learn More
TSPAN16, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TSPAN16.
Brand: Abnova PAB23831.100uL
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein might couple to signal transduction pathways and possibly modulate cellular activation and adhesion in haemopoietic and neural tissue. [provided by RefSeq
Sequence: IVGDVALEHTFVTLRKNYRGYNEPDDYSTQWNLVMEKLKCCGVNNYTDFSGSSFEMTTGHSpecifications
| TSPAN16 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TSPAN16. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| TM-8/TM4-B/TM4SF16 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 26526 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| TSPAN16 | |
| TSPAN16 | |
| Recombinant protein corresponding to amino acids of human TSPAN16. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only