Learn More
TRPV2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TRPV2.
Brand: Abnova PAB28296.100uL
Description
This gene encodes an ion channel that is activated by high temperatures above 52 degrees Celsius. The protein may be involved in transduction of high-temperature heat responses in sensory ganglia. It is thought that in other tissues the channel may be activated by stimuli other than heat. [provided by RefSeq
Sequence: RGKLDFGSGLPPMESQFQGEDRKFAPQIRVNLNYRKGTGASQPDPNRFDRDRLFNAVSRGVPEDLAGLPEYLSKTSKYLTDSSpecifications
| TRPV2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TRPV2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| MGC12549/VRL/VRL-1/VRL1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51393 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| TRPV2 | |
| TRPV2 | |
| Recombinant protein corresponding to amino acids of recombinant TRPV2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only