Learn More
TRIOBP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TRIOBP.
Brand: Abnova PAB20241.100uL
Description
This gene encodes a protein with an N-terminal pleckstrin homology domain and a C-terminal coiled-coil region. The protein interacts with trio, which is involved with neural tissue development and controlling actin cytoskeleton organization, cell motility and cell growth. The protein also associates with F-actin and stabilizes F-actin structures. Mutations in this gene have been associated with a form of autosomal recessive nonsyndromic deafness. Multiple alternatively spliced transcript variants that would encode different isoforms have been found for this gene, however some transcripts may be subject to nonsense-mediated decay (NMD). [provided by RefSeq
Sequence: EIGALMRQAEEREHTLRRCQQEGQELLRHNQELHGRLSEEIDQLRGFIASQGMGNGCGRSNERSSCELEVLLRVKENELQYLKKEVQCLRDELQMMQKDKRFTSGKYQDVYVELSHIKTRSEREIEQLKEHLRLSpecifications
| TRIOBP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TRIOBP. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DFNB28/FLJ39315/HRIHFB2122/KIAA1662/TARA/dJ37E16.4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 11078 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| TRIOBP | |
| TRIOBP | |
| Recombinant protein corresponding to amino acids of human TRIOBP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only