Learn More
TRIM5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRIM5.
Brand: Abnova PAB12911.100ug
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Five alternatively spliced transcript variants for this gene have been described. However, the full length nature of two of the variants has not been determined. [provided by RefSeq
Specifications
| TRIM5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TRIM5. | |
| In PBS (0.02% sodium azide) | |
| RNF88/TRIM5alpha | |
| Rabbit | |
| 100 μg | |
| Primary | |
| A band at approximately 25 KDa can be detected. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| TRIM5 | |
| TRIM5 | |
| A synthetic peptide corresponding to C-terminus 13 amino acids of human TRIM5 delta. | |
| RUO | |
| 85363 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only