Learn More
TRIM41 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TRIM41.
Brand: Abnova PAB21661.100uL
Description
TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 [PubMed 16022281]). See TRIM45 (MIM 609318).[supplied by OMIM
Sequence: RESTHHKEKVGPGGSSVGSGDASSSRHHHRRRRLHLPQQPLLQREVWCVGTNGKRYQAQSSTEQTLLSPSEKPRRFGVYLDYEASpecifications
| TRIM41 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TRIM41. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC1127/MGC31991/RINCK | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 90933 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| TRIM41 | |
| TRIM41 | |
| Recombinant protein corresponding to amino acids of human TRIM41. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only