Learn More
TRIM28, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRIM28.
Brand: Abnova PAB27203.100ug
Description
The protein encoded by this gene mediates transcriptional control by interaction with the Kruppel-associated box repression domain found in many transcription factors. The protein localizes to the nucleus and is thought to associate with specific chromatin regions. The protein is a member of the tripartite motif family. This tripartite motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. [provided by RefSeq
Specifications
| TRIM28 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| TRIM28 | |
| TRIM28 | |
| A synthetic peptide corresponding to 14 amino acids at C-terminus of human TRIM28. | |
| 100 μg | |
| Primary | |
| At least three isoforms of TRIM28 are known to exist; this antibody will detect all three isoforms | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TRIM28. | |
| In PBS (0.02% sodium azide) | |
| FLJ29029/KAP1/RNF96/TF1B/TIF1B | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 10155 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only