Learn More
TRAF3IP2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRAF3IP2.
Brand: Abnova PAB0371.100ug
Description
This gene encodes a protein involved in regulating responses to cytokines by members of the Rel/NF-kappaB transcription factor family. These factors play a central role in innate immunity in response to pathogens, inflammatory signals and stress. This gene product interacts with TRAF proteins (tumor necrosis factor receptor-associated factors) and either I-kappaB kinase or MAP kinase to activate either NF-kappaB or Jun kinase. Several alternative transcripts encoding different isoforms have been identified. Another transcript, which does not encode a protein and is transcribed in the opposite orientation, has been identified. Overexpression of this transcript has been shown to reduce expression of at least one of the protein encoding transcripts, suggesting it has a regulatory role in the expression of this gene. [provided by RefSeq
Sequence: QDLPRPLRSREFPQFEPSpecifications
| TRAF3IP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TRAF3IP2. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| ACT1/C6orf2/C6orf4/C6orf5/C6orf6/CIKS/DKFZp586G0522/MGC3581 | |
| Rabbit | |
| Protein G affinity chromatography | |
| RUO | |
| 10758 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TRAF3IP2 | |
| TRAF3IP2 | |
| A synthetic peptide corresponding to human TRAF3IP2. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only