Learn More
TOMM22, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TOMM22.
Brand: Abnova PAB28636.100uL
Description
The protein encoded by this gene is an integral membrane protein of the mitochondrial outer membrane. The encoded protein interacts with TOMM20 and TOMM40, and forms a complex with several other proteins to import cytosolic preproteins into the mitochondrion. [provided by RefSeq
Sequence: MAAAVAAAGAGEPQSPDELLPKGDAEKPEEELEEDDDEELDETLSERLWGLTEMFPERVRSAAGATFDLSLFVAQKSpecifications
| TOMM22 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TOMM22. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| 1C9-2/MST065/MSTP065/TOM22 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 56993 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| TOMM22 | |
| TOMM22 | |
| Recombinant protein corresponding to amino acids of human TOMM22. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only