Learn More
TNFRSF13B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF13B.
Brand: Abnova PAB12862.100ug
Description
The protein encoded by this gene is a lymphocyte-specific member of the tumor necrosis factor (TNF) receptor superfamily. It interacts with calcium-modulator and cyclophilin ligand (CAML). The protein induces activation of the transcription factors NFAT, AP1, and NF-kappa-B and plays a crucial role in humoral immunity by interacting with a TNF ligand. This gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq
Specifications
| TNFRSF13B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF13B. | |
| In PBS (0.02% sodium azide) | |
| CD267/CVID/FLJ39942/MGC133214/MGC39952/TACI/TNFRSF14B | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TNFRSF13B | |
| TNFRSF13B | |
| A synthetic peptide corresponding to N-terminus of human TNFRSF13B. | |
| RUO | |
| 23495 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only