Learn More
TNFAIP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TNFAIP1.
Brand: Abnova PAB13691.100ug
Description
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner. [provided by RefSeq
Specifications
| TNFAIP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TNFAIP1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| B12/B61/EDP1/MGC2317 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 7126 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunohistochemistry (Frozen sections) (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| TNFAIP1 | |
| TNFAIP1 | |
| A synthetic peptide corresponding to amino acids at N-terminus of human TNFAIP1. | |
| 100 μg | |
| Primary | |
| Identical to the related rat and mouse sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only