missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM135, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TMEM135.
Brand: Abnova PAB27412.100ug
This item is not returnable.
View return policy
Specifications
| TMEM135 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| TMEM135 | |
| TMEM135 | |
| A synthetic peptide corresponding to 16 amino acids at C-terminus of human TMEM135. | |
| 100 μg | |
| Primary | |
| At least two isoforms of TMEM135 are known to exist; this antibody will detect both isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TMEM135. | |
| In PBS (0.02% sodium azide) | |
| DKFZp686I1974/FLJ22104 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 65084 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction