Learn More
TMED10 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TMED10.
Brand: Abnova PAB13235.100ug
Description
This gene is a member of the EMP24/GP25L/p24 family and encodes a protein with a GOLD domain. This type I membrane protein is localized to the plasma membrane and golgi cisternae and is involved in vesicular protein trafficking. The protein is also a member of a heteromeric secretase complex and regulates the complex's gamma-secretase activity without affecting its epsilon-secretase activity. Mutations in this gene have been associated with early-onset familial Alzheimer's disease. This gene has a pseudogene on chromosome 8. [provided by RefSeq]
Specifications
| TMED10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TMED10. | |
| In PBS (0.02% sodium azide) | |
| P24(DELTA)/S31I125/S31III125/TMP21/Tmp-21-I | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| TMED10 | |
| TMED10 | |
| A synthetic peptide corresponding to C-terminus 19 amino acids of human TMED10. | |
| RUO | |
| 10972 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only