Learn More
TMC2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TMC2.
Brand: Abnova PAB24497.100uL
Description
This gene is considered a member of a gene family predicted to encode transmembrane proteins. The specific function of this gene is unknown; however, expression in the inner ear suggests that it may be crucial for normal auditory function. Mutations in this gene may underlie hereditary disorders of balance and hearing. [provided by RefSeq
Sequence: QVLREVEKSHKSVKGKATARDSEDTPKSSSKNATQLQLTKEETTPPSASQSQAMDKKAQGPGTSNSASRTSpecifications
| TMC2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TMC2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| C20orf145/dJ686C3.3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 117532 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| TMC2 | |
| TMC2 | |
| Recombinant protein corresponding to amino acids of human TMC2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only