missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM9SF4 Rabbit anti-Guinea pig, Human, Monkey, Mouse, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TM9SF4.
Brand: Abnova PAB28189.50ug
This item is not returnable.
View return policy
Specifications
| TM9SF4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TM9SF4. | |
| In PBS (0.09% sodium azide) | |
| KIAA0255/dJ836N17.2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 9777 | |
| Guinea Pig, Human, Monkey, Mouse, Rabbit | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15-20 ug/ml) | |
| TM9SF4 | |
| TM9SF4 | |
| A synthetic peptide corresponding to 17 amino acids at internal region of human TM9SF4. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction