missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLX1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TLX1.
Brand: Abnova PAB27431.100ug
This item is not returnable.
View return policy
Specifications
| TLX1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TLX1 | |
| TLX1 | |
| A synthetic peptide corresponding to 16 amino acids at N-terminus of human TLX1. | |
| 100 μg | |
| Primary | |
| Multiple isoforms of TLX1 exist due to alternative splicing events. TLX1 antibody is predicted to not cross-react with other TLX proteins. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TLX1. | |
| In PBS (0.02% sodium azide) | |
| HOX11/MGC163402/TCL3 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 3195 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction