Learn More
TLR8 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TLR8.
Brand: Abnova PAB25701.100ug
Description
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is predominantly expressed in lung and peripheral blood leukocytes, and lies in close proximity to another family member, TLR7, on chromosome X. [provided by RefSeq
Specifications
| TLR8 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TLR8 | |
| TLR8 | |
| A synthetic peptide corresponding to 15 amino acids at C-terminus of human TLR8. | |
| 100 μg | |
| Primary | |
| At least three isoforms of TLR8 are known to exist. TLR8 antibody is predicted to not cross-react with other TLR protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TLR8. | |
| In PBS (0.05% sodium azide) | |
| CD288/MGC119599/MGC119600 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 51311 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only