Learn More
TIMP3 Rabbit anti-Bovine, Dog, Horse, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TIMP3.
Brand: Abnova PAB27956.100ug
Description
This gene belongs to the TIMP gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix (ECM). Expression of this gene is induced in response to mitogenic stimulation and this netrin domain-containing protein is localized to the ECM. Mutations in this gene have been associated with the autosomal dominant disorder Sorsby's fundus dystrophy. [provided by RefSeq]
Specifications
| TIMP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TIMP3. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| HSMRK222/K222/K222TA2/SFD | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 7078 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| TIMP3 | |
| TIMP3 | |
| A synthetic peptide corresponding to amino acids 175-211 of human TIMP3. | |
| 100 μg | |
| Primary | |
| Bovine, Canine, Equine, Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only