Learn More
TIMP3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TIMP3.
Brand: Abnova PAB8014.100ug
Description
This gene belongs to the TIMP gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix (ECM). Expression of this gene is induced in response to mitogenic stimulation and this netrin domain-containing protein is localized to the ECM. Mutations in this gene have been associated with the autosomal dominant disorder Sorsby's fundus dystrophy. [provided by RefSeq]
Sequence: WAPPDKSIINATDPSpecifications
| TIMP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TIMP3. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| HSMRK222/K222/K222TA2/SFD | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 7078 | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TIMP3 | |
| TIMP3 | |
| A synthetic peptide corresponding to amino acids 198-211 at C-terminus of human TIMP3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only