Learn More
THSD1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant THSD1.
Brand: Abnova PAB20606.100uL
Description
The protein encoded by this gene contains a type 1 thrombospondin domain, which is found in a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Alternatively spliced transcript variants encoding different isoforms have been observed for this gene. [provided by RefSeq
Sequence: VPPEDDASGSESFQSNAQKIIPPLFSYRLAQQQLKEMKKKGLTETTKVYHVSQSPLTDTAIDAAPSAPLDLESPEEAAANKFRIKSPFPEQPAVSAGERPPSRLDLNVTQSpecifications
| THSD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant THSD1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC74971/TMTSP/UNQ3010 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55901 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| THSD1 | |
| THSD1 | |
| Recombinant protein corresponding to amino acids of human THSD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only