missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGM5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TGM5.
Brand: Abnova PAB25735.100ug
This item is not returnable.
View return policy
Specifications
| TGM5 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TGM5 | |
| TGM5 | |
| A synthetic peptide corresponding to 19 amino acids at C-terminus of human TGM5. | |
| 100 μg | |
| Primary | |
| TGM5 antibody is predicted to not cross-react with other TGase protein family members. Multiple isoforms of TGM5 are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TGM5. | |
| In PBS (0.02% sodium azide) | |
| MGC141907/TGM6/TGMX/TGX | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 9333 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction