Learn More
TGFBR1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TGFBR1.
Brand: Abnova PAB3503.400uL
Description
The protein encoded by this gene forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS). Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| TGFBR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TGFBR1. | |
| In PBS (0.09% sodium azide) | |
| AAT5/ACVRLK4/ALK-5/ALK5/LDS1A/LDS2A/SKR4/TGFR-1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 7046 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| TGFBR1 | |
| TGFBR1 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human TGFBR1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only