missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTN3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of TCTN3.
Brand: Abnova PAB19274.100ug
This item is not returnable.
View return policy
Specifications
| TCTN3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (4 ug/mL) The optimal working dilution should be determined by the end user. | |
| TCTN3 | |
| TCTN3 | |
| A synthetic peptide corresponding to 17 amino acids near C-terminus of human TCTN3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of TCTN3. | |
| In PBS (0.02% sodium azide) | |
| C10orf61/DKFZp564D116/TECT3 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 26123 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction