Learn More
TCF21 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TCF21.
Brand: Abnova PAB21931.100uL
Description
TCF21 encodes a transcription factor of the basic helix-loop-helix family. The TCF21 product is mesoderm specific, and expressed in embryonic epicardium, mesenchyme-derived tissues of lung, gut, gonad, and both mesenchymal and glomerular epithelial cells in the kidney. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: LSDVEDLQEVEMLECDGLKMDSNKEFVTSNESTEESSNCENGSPQKGRGGLGKRRKAPTKKSPLSGVSQEGKQVSpecifications
| TCF21 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TCF21. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| POD1/bHLHa23 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6943 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| TCF21 | |
| TCF21 | |
| Recombinant protein corresponding to amino acids of human TCF21. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only