Learn More
SYT2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of SYT2.
Brand: Abnova PAB25436.100ug
Description
Synaptotagmins, like SYT2, are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis (Hilbush and Morgan, 1994 [PubMed 8058779]).[supplied by OMIM
Sequence: R-K-Tp-L-NSpecifications
| SYT2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of SYT2. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| SytII | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 127833 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence | |
| Unconjugated | |
| Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| SYT2 | |
| SYT2 | |
| Synthetic phosphopeptide corresponding to residues surrounding T202 of human SYT2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only