missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYN1 (phospho S9), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Syn1.
Brand: Abnova PAB9664.100uL
This item is not returnable.
View return policy
Specifications
| SYN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of Syn1. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| Syn1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S9 of rat Syn1. | |
| 100 μL | |
| Primary | |
| This antibody also weakly labels the ∼55k synapsin II protein which has a similar phosphorylation site to that of Ser9 on synapsin I. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Syn1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 24949 | |
| Bovine, Frog, Human, Mouse, Rat, Zebrafish | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction