missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYCP3 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Sycp3.
Brand: Abnova PAB12013.100uL
This item is not returnable.
View return policy
Specifications
| SYCP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Sycp3. | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| Cor1/Scp3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to mouse SCP3. | |
| Store at 4°C. Do not freeze. |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:750) The optimal working dilution should be determined by the end user. | |
| Sycp3 | |
| Sycp3 | |
| A synthetic peptide corresponding to N-terminus of mouse Sycp3. | |
| RUO | |
| 20962 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction