Learn More
SUPT7L, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SUPT7L.
Brand: Abnova PAB23560.100uL
Description
SUPT7L is a protein subunit of the human STAGA complex (SPT3; (MIM 602947)/TAF9 (MIM 600822)/GCN5 (MIM 602301) acetyltransferase complex), which is a chromatin-modifying multiprotein complex (Martinez et al., 2001 [PubMed 11564863]).[supplied by OMIM
Sequence: HEYCLKFTKLLRFAVDREARLGQTPFPDVMEQVFHEVGIGSVLSLQKFWQHRIKDYHSYMLQISKQLSEEYERIVNPEKATEDAKPVKIKSpecifications
| SUPT7L | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SUPT7L. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| ART1/KIAA0764/MGC90306/SPT7L/STAF65/STAF65(gamma) | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9913 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| SUPT7L | |
| SUPT7L | |
| Recombinant protein corresponding to amino acids of human SUPT7L. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only