Learn More
SULT2A1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SULT2A1.
Brand: Abnova PAB2528.400uL
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene is primarily expressed in liver and adrenal tissues where the encoded protein sulfates steroids and bile acids. [provided by RefSeq
Specifications
| SULT2A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SULT2A1. | |
| In PBS (0.09% sodium azide) | |
| DHEA-ST/DHEAS/HST/ST2/ST2A3/STD/hSTa | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 6822 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SULT2A1 | |
| SULT2A1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human SULT2A1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only