Learn More
SULT1B1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SULT1B1.
Brand: Abnova PAB28569.100uL
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes. [provided by RefSeq
Sequence: KIEQFHSRPDDIVIATYPKSGTTWVSEIIDMILNDGDIEKCKRGFITEKVPMLEMTLPGLRTSGIEQLEKNPSPRIVKTHLPTDLLPKSFWENNCKMIYLARNSpecifications
| SULT1B1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SULT1B1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| MGC13356/ST1B2/SULT1B2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 27284 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| SULT1B1 | |
| SULT1B1 | |
| Recombinant protein corresponding to human SULT1B1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only