missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulf1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a recombinant Sulf1.
Brand: Abnova PAB8523.50ug
This item is not returnable.
View return policy
Specifications
| Sulf1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a recombinant Sulf1. | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| Sulf1 | |
| Recombinant protein corresponding to rat Sulf1. | |
| RUO | |
| 171396 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Sulf1 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction