Learn More
STON2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant STON2.
Brand: Abnova PAB20161.100uL
Description
Endocytosis of cell surface proteins requires a dynamic complex of proteins that assemble on the inner surface of the plasma membrane. Stonin-2 is a component of the endocytic machinery that likely regulates vesicle endocytosis.[supplied by OMIM
Sequence: RTPSVTEAPPWRATNPFLNETLQDVQPSPINPFSAFFEEQERRSQNSSISSTTGKSQRDSLIVIYQDAISFDDSSKTQSHSDAVEKLKQLQIDDPDHFGSATLPDDDPVAWIELDAHPPGSARSQPRDGWPMMLRIPEKSpecifications
| STON2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant STON2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| STN2/STNB/STNB2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 85439 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| STON2 | |
| STON2 | |
| Recombinant protein corresponding to amino acids of human STON2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only