Learn More
STK39 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STK39.
Brand: Abnova PAB16586.100ug
Description
This gene encodes a serine/threonine kinase that is thought to function in the cellular stress response pathway. The kinase is activated in response to hypotonic stress, leading to phosphorylation of several cation-chloride-coupled cotransporters. The catalytically active kinase specifically activates the p38 MAP kinase pathway, and its interaction with p38 decreases upon cellular stress, suggesting that this kinase may serve as an intermediate in the response to cellular stress. [provided by RefSeq
Specifications
| STK39 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of STK39. | |
| In PBS (0.02% sodium azide) | |
| DCHT/DKFZp686K05124/PASK/SPAK | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| STK39 | |
| STK39 | |
| A synthetic peptide corresponding to internal region 18 amino acids of human STK39. | |
| RUO | |
| 27347 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only